Order Entry
Spain
ContactUsLinkComponent
8020A Calibration mix (10 components)
8020A Calibration mix (10 components)
Catalog # REST30222
Supplier:  Restek
CAS Number:  
undefined
8020A Calibration mix (10 components)
Catalog # REST30222
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml each in P and T methanol
  • Pk:
    1
  • Description:
    Calibration mix, 8020A
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Mix contains: Benzene (71-43-2); Chlorobenzene (108-90-7); 1,2-Dichlorobenzene (95-50-1); 1,3-Dichlorobenzene (541-73-1); 1,4-Dichlorobenzene (106-46-7); Ethylbenzene (100-41-4); Toluene (108-88-3); m-Xylene (108-38-3); o-Xylene (95-47-6); p-Xylene (106-42-3).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.