Order Entry
Spain
ContactUsLinkComponent
SV calibration mix #2 (15 components)
SV calibration mix #2 (15 components)
Catalog # REST31008
Supplier:  Restek
CAS Number:  
undefined
SV calibration mix #2 (15 components)
Catalog # REST31008
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml each in methylene chloride
  • Pk:
    1
  • Description:
    SV calibration mix #2
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

SV calibration mix #2 contains benzoic acid, 4-chloro-3-methylphenol, 2-chlorophenol, 2,4-dichlorophenol, 2,4-dimethylphenol, 4,6-dinitro-2-methylphenol (Dinitro-o-cresol), 2,4-dinitrophenol, 2-methylphenol (o-cresol), 4-methylphenol (p-cresol), 2-nitrophenol, 4-nitrophenol, pentachlorophenol, phenol, 2,4,5-trichlorophenol, 2,4,6-trichlorophenol.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.