Order Entry
Spain
ContactUsLinkComponent
SV calibration mix #3 (14 components)
SV calibration mix #3 (14 components)
Catalog # REST31009
Supplier:  Restek
CAS Number:  
undefined
SV calibration mix #3 (14 components)
Catalog # REST31009
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml each in methylene chloride
  • Pk:
    1 mL
  • Description:
    SV calibration mix #3
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

SV calibration mix contains bis(2-chloroethoxy)methane, bis(2-chloroethyl)ether, bis(2-ethylhexyl)phthalate, 4-bromophenyl phenyl ether, benzyl butyl phthalate, 2-chloronaphthalene, 4-chlorophenyl phenyl ether, dimethylphthalate, di-n-butylphthalate, di-n-octylphthalate, N-nitrosodimethylamine, N-nitroso-di-n-propylamine, N-nitrosodiphenylamine, 2,2'-oxybis(1-chloropropane).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.