Herbicide reference standard, 2,4-dichlorophenyl acetic acid methyl ester (DCAA methyl ester), derivatised form
Catalog # REST32050
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 µg/ml in hexane
- Pk:1 mL
- CAS No.:55954-23-9
- Description:2,4-dichlorophenyl acetic acid methyl ester (DCAA methyl ester)
- Formula:C₉H₈Cl₂O₂
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.