Order Entry
Spain
ContactUsLinkComponent
TNRCC 1006 retention time marker mix (9 components)
TNRCC 1006 retention time marker mix (9 components)
Catalog # REST31814
Supplier:  Restek
CAS Number:  
undefined
TNRCC 1006 retention time marker mix (9 components)
Catalog # REST31814
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    200 µg/ml in pentane
  • Pk:
    1 Ampoul
  • Description:
    TNRCC 1006 retention time marker mix
  • Pk Info:
    1 ml/ ampoule

Specifications

About this item

This TNRCC 1006 retention time marker mix contains: (C6) n-Hexane, (C7) n-Heptane, (C8) n-Octane, (C10) n-Decane, (C12) n-Dodecane, (C16) n-Hexadecane, (C21) n-Heneicosane, (C28) n-Octacosane, (C35) n-Pentatriacontane.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.