608 calibration mix (16 components)
REST32022
: Restek
:
- Concentration:200 μg/ml each in hexane:toluene (1:1)
- Pk:1 mL
- Description:608 calibration mix
- Pk Info:1 ml/ampoule
This mix contains 16 components.
Components are Aldrin, α-BHC, β-BHC, δ-BHC, γ-BHC (Lindane), 4,4'-DDD, 4,4'-DDE, 4,4'-DDT, dieldrin, endosulphan I, endosulphan II, endosulphan sulphate, endrin, endrin aldehyde, heptachlor, heptachlor epoxide (isomer B).
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.