Order Entry
Spain
ContactUsLinkComponent
Herbicide reference standard, 2,4,5-TP (Silvex) methyl ester
Herbicide reference standard, 2,4,5-TP (Silvex) methyl ester
Catálogo # REST32246
Supplier:  Restek
CAS Number:  
undefined
Herbicide reference standard, 2,4,5-TP (Silvex) methyl ester
Catálogo # REST32246
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    1000 μg/ml in methanol
  • Pk:
    1
  • CAS No.:
    4841-20-7
  • Description:
    2,4,5-TP (Silvex) methyl ester
  • Formula:
    C₁₀H₉Cl₃O₃
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.