2-Propanol
2-Propanol, 50,000 µg/ml in water, 1 ml/ampoule, 1 mL
Catalog # REST30473
Supplier: Restek
Jump to:
Product Details & Documents
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Specifications
Specifications
- Concentration:50,000 µg/ml in water
- Pk:1 mL
- Description:2-Propanol
- Pk Info:1 ml/ampoule
- Cat. No.:REST30473
- Description:2-Propanol
- Melting point:C₃H₈O
- CAS No.:67-63-0



