1-Butanol
1-Butanol, 50000 µg/ml in P&T methanol, 1 ml/ampoule, 1 mL
Catalog # REST30474
Supplier: Restek
Jump to:
Product Details & Documents
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
- Concentration:50000 µg/ml in P&T methanol
- Pk:1 mL
- Description:1-Butanol
- Pk Info:1 ml/ampoule
- Cat. No.:REST30474
- Formula:C₄H₁₀O
- Description:1-Butanol
- CAS No.:71-36-3



