Pesticide reference standard, hexachlorocyclopentadiene
Catalog # REST32232
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 μg/ml in methanol
- Pack type:Ampoule
- Pk:1
- CAS No.:77-47-4
- Description:Hexachlorocyclopentadiene
- Formula:C₅Cl₆
- Description:Hexachlorocyclopentadiene
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.