Order Entry
Spain
Orders LinkContactUsLinkComponent
 

δ-BHC

δ-BHC, 1,000 µg/ml in methanol, 1 ml/ampoule, 1
  REST32217
 :   Restek
δ-BHC
  REST32217
 :  Restek
 :  
δ-BHC
δ-BHC

δ-BHC, 1,000 µg/ml in methanol, 1 ml/ampoule, 1

 :  
 
55,00 €
   
1 * 1 UN
 

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

 

 

  • Concentration:
    1,000 µg/ml in methanol
  • Pk:
    1
  • Description:
    δ-BHC
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST32217
  • Formula:
    C₆H₆Cl₆
  • Description:
    δ-BHC
  • CAS No.:
    319-86-8

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.