Order Entry
Spain
ContactUsLinkComponent
 
Pesticide reference Standard, Heptachlor epoxide (isomer B)
  REST32230
 :  Restek
 :  
undefined
Pesticide reference Standard, Heptachlor epoxide (isomer B)
  REST32230
 :  Restek
 :  

 

  • Concentration:
    1000 μg/ml in methanol
  • Pack type:
    Ampoule
  • Pk:
    1
  • CAS No.:
    1024-57-3
  • Description:
    Heptachlor epoxide (isomer B)
  • Formula:
    C₁₀H₅Cl₇O
  • Storage temperature:
    2…8 °C
  • Description:
    Heptachlor epoxide (isomer B)
  • Pk Info:
    1 ml/ampoule

 

 

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.