Residual range calibration standard (RCS)
Catalog # REST31817
Supplier: Restek
CAS Number:
Specifications
- Concentration:50000 µg/ml in methylene chloride
- Pk:1 Ampoul
- Description:Residual range calibration standard (RCS)
- Pk Info:1 ml
Specifications
About this item
SAE30 motor oil:SAE40 motor oil (1:1).
- 2 components