Order Entry
Spain
ContactUsLinkComponent
DHA PONA standard (188 components)
DHA PONA standard (188 components)
Catalog # REST30731
Supplier:  Restek
CAS Number:  
undefined
DHA PONA standard (188 components)
Catalog # REST30731
Supplier:  Restek
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    DHA PONA standard, neat
  • Pk Info:
    0,15 ml in autosampler vial

Specifications

About this item

This standard is a qualitative mixture of various gasoline and refinery materials prepared to provide nearly every component that may be encountered in feedstock and finished gasolines. Some oxygenates have been added to allow this blend to be used by refineries for detailed hydrocarbon analysis (DHA) method setup and to fulfill requirements.

Contains 188 components.

Certifications: Fulfill requirements such as ASTM Methods D6729, D6730, and D6733.