Picloram, reference standard
Picloram, 1000 µg/ml in methanol, 1 ml/ampoule, 1
Catalog # REST32265
Supplier: Restek
Jump to:
Product Details & Documents
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
- Pk:1
- Description:Picloram, 1000 µg/ml in methanol
- Pk Info:1 ml/ampoule
- Cat. No.:REST32265
- Formula:C₆H₃Cl₃N₂O₂
- Description:Picloram
- CAS No.:1918-02-1



