Order Entry
Spain
Orders LinkContactUsLinkComponent
Aldrin, reference standard

Aldrin, reference standard

Aldrin, 1000 µg/ml in methanol, 1 ml/ampoule, 1
Catalog # REST32205
Supplier:   Restek
Aldrin, reference standard
Catalog # REST32205
Supplier:  Restek
CAS Number:  
Aldrin, reference standard

Aldrin, 1000 µg/ml in methanol, 1 ml/ampoule, 1

CAS Number:  
Price
53,00 €
Log in for availability
1 * 1 UN
Product Details & Documents

Certificaciones: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

Specifications

Specifications

  • Pk:
    1
  • Description:
    Aldrin, 1000 µg/ml in methanol
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST32205
  • Formula:
    C₁₂H₈Cl₆
  • Description:
    Aldrin
  • CAS No.:
    309-00-2

Specifications