Chromium(III) nitrate nonahydrate ≥98.0%, Puriss. p.a., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7789-02-8
:
Formula:CrN₃O₉·9H₂O
MDL Number:MFCD00149659
:400.15 g/mol
Merck Index:13,02246
Boiling Pt: Dec. (on heating)
Melting Pt:54.8 °C
Density:1.89 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3260
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Assay:98.0 - 102.0 %
- Calcium (Ca):Max. 50 ppm
- Cadmium (Cd):Max. 10 ppm
- Copper (Cu):Max. 10 ppm
- Iron (Fe):Max. 200 ppm
- Potassium (K):Max. 50 ppm
- Sodium (Na):Max. 50 ppm
- Nickel (Ni):Max. 50 ppm
- Lead (Pb):Max. 20 ppm
- Zinc (Zn):Max. 10 ppm
- Total S (as SO₄):Max. 50 ppm
- Chloride (Cl):Max. 50 ppm
- Ammonium (NH₄):Max. 10 ppm
Specification Test Results
Frequently Bought Together