Order Entry
Spain
ContactUsLinkComponent
 
Dimethyl sulphoxide ≥99.9% ACS, Riedel-de Haën™
    
 
undefined
Dimethyl sulphoxide ≥99.9% ACS, Riedel-de Haën™
    

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

 

 

  • Appearance:
    complying
  • Assay (GC):
    Min. 99.90 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Non-volatile matter:
    Max. 0.01 %
  • Acid (by titration):
    Max. 0.001 meq/g
  • IR spectrum:
    complying
  • Refractive Index:
    n20/D 1.479 (lit.)

 

 

Frequently Bought Together