Order Entry
Spain
ContactUsLinkComponent
 
Dimethyl sulphoxide ≥99.8% (by GC) for UV spectrophotometry, Riedel-de Haën™
Catalog # 41641-250ML
   Safer Human and Environmental Health
CAS Number:  
undefined
Dimethyl sulphoxide ≥99.8% (by GC) for UV spectrophotometry, Riedel-de Haën™
Catalog # 41641-250ML
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Vapor Density:
    2.7 (vs air)
  • Assay (GC):
    Min. 99.8 %
  • Refractive index (n 20/D):
    1.478 - 1.480
  • Non-volatile matter:
    Max. 0.002 %
  • Water (Karl Fischer):
    Max. 0.2 %
  • Absorbance at 360 nm:
    Max. 0.003
  • Absorbance at 340 nm:
    Max. 0.01
  • Absorbance at 320 nm:
    Max. 0.04
  • Absorbance at 300 nm:
    Max. 0.10
  • Absorbance at 290 nm:
    Max. 0.16
  • Absorbance 280 nm:
    Max. 0.22
  • Absorbance at 270 nm:
    Max. 0.4
  • Absorbance at 265 nm:
    Max. 0.8

Specification Test Results

About this item

 :
 :78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C
Melting Pt:18.5 °C
Density:1.104 g/cm³ (20 °C)
Flash Pt:28.5 %(V)
Storage Temperature:Ambient
EINECS:200-664-3
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089