Order Entry
Spain
ContactUsLinkComponent
 
 
 
Chloroform ≥99.5% stabilised for IR spectrophotometry, Riedel-de Haën™
  25670-500ML
 :  
undefined
 
 
Chloroform ≥99.5% stabilised for IR spectrophotometry, Riedel-de Haën™
  25670-500ML
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • IR spectrum:
    complying
  • Purity excluding stabilizer (GC):
    Min. 99.5 %
  • Refractive index (n 20/D):
    1.445 - 1.447
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Ethanol (GC):
    0.2 - 1.0 Gew.%
  • Vapor Pressure:
    160 mmHg ( 20 °C)
  • Vapor Density:
    4.1 (vs air)
  • Refractive Index:
    n20/D 1.446
  • Contains:
    ~0.5% ethanol as stabilizer

 

 

 :
 :119.38 g/mol
 :13,02160
 :60…62 °C
 :-63 °C
 :–63 °C
 :1.48 g/cm³
 :Ambient
 :200-663-8
 :1888
 :6.1,III
 :Stabilised with ethanol ~0.5%
 :05-2114614906-45
 :GHS08
 :GHS06
 :67-66-3
 :CHCl₃
 :MFCD00000826