


Sodium perchlorate monohydrate ≥98.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7791-07-3
:
Formula:NaClO₄·H₂O
MDL Number:MFCD00149164
:140.46 g/mol
Merck Index:12,08798
Boiling Pt: Dec. (on heating)
Melting Pt:469 °C (anhydr.)
Density:2.02 g/cm³ (20 °C)
UN:1502
ADR:5.1,II
:GHS07
:GHS08
:GHS03
Storage Temperature:Ambient
Specification Test Results
- Assay:98.0 - 102.0 %
- Assay of NaClO4:85.0 - 90.0 %
- Solution 5% in H₂O:complying
- pH (5 %, 25 °C):5.0 - 8.0
- Insoluble matter:Max. 0.005 %
- Heavy metals (as Pb):Max. 5 ppm
- Calcium (Ca):Max. 10 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 3 ppm
- Potassium (K):Max. 50 ppm
- Magnesium (Mg):Max. 5 ppm
- Manganese (Mn):Max. 5 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Zinc (Zn):Max. 5 ppm
- Sulfate (SO₄):Max. 20 ppm
- Total nitrogen (as N):Max. 5 ppm
- Chloride (Cl):Max. 30 ppm
Specification Test Results
Frequently Bought Together