Mouse Recombinant FGF-9 (Animal free) (from E. coli)
Interferon gamma (IFN-γ) is a type II interferon that is critical during adaptive and innate immune responses to infection. IFN-γ is produced by T cells and natural killer cells following antigen-specific activation. IFN-γ binds IFN-γ receptors (IFN-γ R1 and IFN-γ R2), which are expressed on most immune cells, to activate the JAK-STAT pathway. IFN-γ-induced signaling increases the expression of class 1 major histocompatibility complex (MHC) molecules. Mouse IFN-γ is not cross-reactive with human IFN-γ.
- High quality
- Low endotoxin
- Affordable
: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
: For research use only.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤10 ng/ml, bioactivity ≥1,0×10⁵ units/mg
- Gene ID:P54130
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:Heparin-binding growth factor -9 (HBGF-9)|Glia activating factor (GAF)
- Protein/peptide name:FGF9
- Purity:≥95%
- Molecular weight:23,4 kDa
- Sequence:MPLGEVGSYF GVQDAVPFGN VPVLPVDSPV LLNDHLGQSE AGGLPRGPAV TDLDHLKGIL RRRQLYCRTG FHLEIFPNGT IQGTRKDHSR FGILEFISIA VGLVSIRGVD SGLYLGMNEK GELYGSEKLT QECVFREQFE ENWYNTYSSN LYKHVDTGRR YYVALNKDGT PREGTRTKRH QKFTHFLPRP VDPDKVPELY KDILSQS
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Frequently Bought Together