Order Entry
Spain
ContactUsLinkComponent
 
PIPPS (1,4-Piperazinedipropanesulphonic acid) ≥97% (by titration analysis on dried basis), Calbiochem®
Catalog # 528315-100
CAS Number:  
PIPPS (1,4-Piperazinedipropanesulphonic acid) ≥97% (by titration analysis on dried basis), Calbiochem®
Catalog # 528315-100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :130.093 g/mol
Storage Temperature:Refrigerator
CAS Number:5625-56-9
Formula:C₁₀H₂₂N₂O₆S₂
MDL Number:MFCD00274122