Tubing connectors for threaded connection caps
Supplier: LENZ, E. (Lenz Laborglas GmbH & CO.KG)
Certificates
About this item
PP with silicone seals.
- Suitable for threaded connection caps with GL 14 and GL 18 threads
- Robust
Ordering information: Caps can be ordered separately.
Frequently Bought Together