Order Entry
Spain
ContactUsLinkComponent
L(+)-Glutamic acid 98.5-101.5% (dry basis), Ultrapure
L(+)-Glutamic acid 98.5-101.5% (dry basis), Ultrapure
Certificates
undefined
L(+)-Glutamic acid 98.5-101.5% (dry basis), Ultrapure

Specification Test Results

  • Assay (Unspecified) - Dry Basis:
    98.5 - 101.5%
  • Identification (FTIR):
    Conforms
  • Loss on Drying:
    ≤0.1%
  • Optical Rotation (c=10 in 2 N HCl):
    31.2° - 32.2°
  • Arsenic (As):
    ≤1.5 ppm
  • Heavy Metals (Pb):
    ≤1.5 ppm
  • Lead (Pb):
    ≤10 ppm

Specification Test Results

Related Information

Frequently Bought Together