Calibration mixture, Connecticut ETPH
Catalog # REST31614
Supplier: Restek
CAS Number:
Specifications
- Pack type:Ampoule
- Pk:1 mL
- Description:Connecticut ETPH calibration mixture
Specifications
About this item
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs.
- 15 components- all n- alkanes