Order Entry
Spain
ContactUsLinkComponent
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid ≥99.0% (by HPLC), white to off-white powder, Novabiochem®
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid ≥99.0% (by HPLC), white to off-white powder, Novabiochem®
Certificates
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid ≥99.0% (by HPLC), white to off-white powder, Novabiochem®

Specification Test Results

  • Colour Index (0.5 M in DMF):
    ≤150 Hazen
  • Identity (IR):
    Passes Test
  • Enantiomeric Purity:
    ≥99.8% (a/a)
  • Purity (HPLC):
    ≥99.0% (a/a)
  • Fmoc-ß-Ala-OH (HPLC):
    ≤0.1% (a/a)
  • Fmoc-ß-Ala-Asp (OtBu)-OH (HPLC):
    ≤0.1% (a/a)
  • Fmoc-Asp(OtBu)-Asp(OtBu)-OH (HPLC):
    ≤0.1% (a/a)
  • Fmoc-Asp-OH (HPLC):
    ≤0.1% (a/a)
  • Assay Free Amino Acid (GC):
    ≤0.2%
  • Purity (TLC(011A)):
    ≥98%
  • Purity (TLC(0811)):
    ≥98%
  • Solubility (25 mmole in 50 ml DMF):
    Clearly Soluble
  • Assay (Acidimetric):
    ≥98.0%
  • Water (K. F.):
    ≤2.0%
  • Ethyl Acetate (HS-GC):
    ≤0.5%
  • Acetate (IC):
    ≤0.02%

Specification Test Results

Frequently Bought Together