Order Entry
Spain
ContactUsLinkComponent
VWR® PerfectBlue Mini L Revolution, Gel System
VWR® PerfectBlue Mini L Revolution, Gel System
Catalog # 700-0799
Supplier:  VWR Collection
undefined
VWR® PerfectBlue Mini L Revolution, Gel System
Catalog # 700-0799
Supplier:  VWR Collection

Specifications

  • Description:
    PerfectBlue gel system Mini L 'Revolution'
  • Seq:
    01
  • Model:
    Mini L Revolution
  • Gel volume:
    85 ml
  • Buffer volume:
    ~800 ml
  • Run time:
    ≥60 h
  • Gel size W×L:
    120×140 mm
  • Sample capacity:
    8 – 100
  • Unit W×L×H:
    170×250×95 mm
  • Pk:
    1

Specifications

About this item

The 'Revolution' series includes a buffer circulation system, powered by the electrodes themselves. Hydrogen bubbles produced at the cathode pass up a diagonally positioned pipe connected to the two sides of the tank producing a gentle circulation of the buffer. The result is no pH and ionic gradient – without need of mechanical pumps.

*Based on an agarose gel of approximately 5 mm thickness

Ordering information: System is supplied with 2 combs (12 and 20 teeth, 1,5 mm thickness) as standard.

Combs

Product Information