Order Entry
Spain
ContactUsLinkComponent
Hexamethylenetetramine 99%
Irritant
Flammable
Hexamethylenetetramine 99%
Certificates
undefined
Irritant
Flammable
Hexamethylenetetramine 99%
Certificates

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Ash:
    ≤0.1 %
  • Titration with HClO4:
    ≥98.5 %
  • NMR (1H-NMR):
    Consistent with structure
  • Particle size:
    ≥45 % (> 75 µm)
  • Residue after ignition:
    ≤0.05 %
  • pH:
    5 to 7

Specification Test Results

Related Information

Frequently Bought Together