Human Recombinant IL-9 (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Interleukin 9 (IL-9) is a cytokine produced by type 2 T helper (Th2) cells and regulates hematopoietic cells. IL-9 signals through the interleukin 9 receptor (IL9R) to activate STAT signaling. IL-9 functions to induce cell proliferation, prevent cell apoptosis, and is associated with asthma and airway hyperresponsiveness.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity assay: MO7e cell proliferation, ED50 ≤2 ng/ml, Bioactivity ≥5,0×10⁵ units/mg
- Gene ID:P15248
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:T-cell growth factor p40|P40 cytokine
- Protein/peptide name:IL-9
- Purity:≥95%
- Molecular weight:14,3 kDa
- Sequence:MQGCPTLAGI LDINFLINKM QEDPASKCHC SANVTSCLCL GIPSDNCTRP CFSERLSQMT NTTMQTRYPL IFSRVKKSVE VLKNNKCPYF SCEQPCNQTT AGNALTFLKS LLEIFQKEKM RGMRGKI
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together