Rat Recombinant MIP-1 A (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Interleukin 17F (IL-17F), a member of the IL-17 cytokine family, is secreted by activated CD4+ T cells and monocytes. IL-17F binds the IL-17 receptor related molecule, IL17RC, to promote the production of the interleukin 6 (IL-6), interleukin 8 (IL-8), and granulocyte macrophage colony-stimulating factor (GM-CSF) cytokines.
- High quality
- Low endotoxin
- Affordable
IL-17F also functions to regulate matrix turnover rates, inhibit endothelial cell angiogenesis, and induce the endothelial cell expression of interleukin 2 (IL-2), monocyte chemoattractant protein-1 (MCP-1), and transforming growth factor beta 1 (TGF-β1).
Delivery: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P50229
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CCL3|LD78α
- UniProtKB:P50229
- Protein/peptide name:MIP-1alpha
- Purity:≥95%
- Molecular weight:7,9 kDa
- Sequence:APYGADTPTA CCFSYGRQIP RKFIADYFET SSLCSQPGVI FLTKRNRQIC ADPKETWVQE YITELELNA
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together