Order Entry
Spain
ContactUsLinkComponent
Human Recombinant MYOSTATIN (from E. coli)
Human Recombinant MYOSTATIN (from E. coli)
Certificates
undefined
Human Recombinant MYOSTATIN (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: MPC-11 cell cytotoxicity, ED50 ≤50 ng/ml, bioactivity ≥2,0×10⁴ units/mg
  • Gene ID:
    O14793
  • Reconstitution instructions:
    Sterile 20 mM HCl at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    GDF-8|MSTN
  • UniProtKB:
    O14793(267-375)
  • Protein/peptide name:
    Myostatin
  • Purity:
    ≥95%
  • Molecular weight:
    12,4/24,8 kDa
  • Sequence:
    DFGLDCDEHS TESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGECEFV FLQKYPHTHL VHQANPRGSA GPCCTPTKMS PINMLYFNGK EQIIYGKIPA MVVDRCGCS
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Related Information

Frequently Bought Together