Order Entry
Spain
ContactUsLinkComponent
Human Recombinant CTGF (from E. coli)
Human Recombinant CTGF (from E. coli)
Certificates
undefined
Human Recombinant CTGF (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: HUVEC cell proliferation, ED50 ≤2000 ng/ml, bioactivity ≥5,0×10^2 units/mg
  • Gene ID:
    P29279
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    HCS24|CCN2
  • UniProtKB:
    P29279
  • Protein/peptide name:
    CTGF
  • Purity:
    ≥90%
  • Molecular weight:
    11,2 kDa
  • Sequence:
     MGKKCIRTPK ISKPIKFELS GCTSMKTYRA KFCGVCTDGR CCTPHRTTTL PVEFKCPDGE VMKKNMMFIK TCACHYNCPG DNDIFESLYY RKMYGDMA
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together