Macrophage inflammatory protein-3 beta (MIP-3 β), also called CCL19, is a chemokine that is expressed in the thymus, lymph nodes, and activated bone marrow stromal cells. MIP-3 β signals through the G protein-coupled receptor CCR7 to regulate normal lymphocyte recirculation. MIP-3 β also functions during T cell trafficking to the thymus, and in T cell and B cell homing to the lymph nodes and secondary lymphoid organs. Human MIP-3 β shows activity on mouse cells.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q99731
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CCL19|SCYA19|ELC|exodus-3
- UniProtKB:Q99731
- Protein/peptide name:MIP-3 beta
- Purity:≥95%
- Molecular weight:8,8 kDa
- Sequence:GTNDAEDCCL SVTQKPIPGY IVRNFHYLLI KDGCRVPAVV FTTLRGRQLC APPDQPWVER IIQRLQRTSA KMKRRSS
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT