
6-Chloro-2-hydroxypyridine ≥98.0% (by GC, titration analysis)
Catalog # TCIAC1079-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:125.0 to 132.0 deg-C
Specification Test Results
About this item
:
:129.55 g/mol
Melting Pt:128.5…129 °C
EINECS:240-909-1
:GHS07
CAS Number:16879-02-0
Formula:C₅H₄ClNO
MDL Number:MFCD00006278