Order Entry
Spain
ContactUsLinkComponent
 
3-Amino-2-chloropyridine 96%
 
undefined
3-Amino-2-chloropyridine 96%

 

  • Appearance:
    Off-white to yellow to pink Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    76 to 81 °C
  • HPLC:
    ≥95 %

 

 

Frequently Bought Together