Mouse Recombinant IL-12 (from HEK293 Cells)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Interleukin 12 (IL-12) is an inducer of cell-mediated immunity and is produced by dendritic cells, macrophages, and B cells after antigen stimulation. IL-12 functions to promote naïve T cell differentiation into the type 1 T helper cell (Th1) lineage. IL-12 also stimulates the production of interferon gamma (IFN-γ) and tumor necrosis factor alpha (TNF-α) from T cells and natural killer cells. Mouse IL-12 is active on human and mouse cells.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity assay: Induces secretion of SEAP in HEK-BlueTM IL-12 cells, ED50 ≤15 ng/ml, Bioactivity ≥6,7×10⁴ units/mg
- Gene ID:P43432/P43431
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:NKSF|CLMF
- Protein/peptide name:IL-12
- Purity:≥95%
- Molecular weight:p35: 21,7 kDa, p40: 35,8 kDa, total: 57,5 kDa
- Sequence:p35: RVIPVSGPAR CLSQSRNLLK TTDDMVKTAR EKLKHYSCTA EDIDHEDITR DQTSTLKTCL PLELHKNESC LATRETSSTT RGSCLPPQKT SLMMTLCLGS IYEDLKMYQT EFQAINAALQ NHNHQQIILD KGMLVAIDEL MQSLNHNGET LRQKPPVGEA DPYRVKMKLC ILLHAFSTRV VTINRVMGYL SSA
p40: MWELEKDVYV VEVDWTPDAP GETVNLTCDT PEEDDITWTS DQRHGVIGSG KTLTITVKEF LDAGQYTCHK GGETLSHSHL LLHKKENGIW STEILKNFKN KTFLKCEAPN YSGRFTCSWL VQRNMDLKFN IKSSSSSPDS RAVTCGMASL SAEKVTLDQR DYEKYSVSCQ EDVTCPTAEE TLPIELALEA RQQNKYENYS TSFFIRDIIK PDPPKNLQMK PLKNSQVEVS WEYPDSWSTP HSYFSLKFFV RIQRKKEKMK ETEEGCNQKG AFLVEKTSTE VQCKGGNVCV QAQDRYYNSS CSKWACVPCR VRS - Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together