Human Recombinant TGF-A (from E. coli)
Proveedor: Shenandoah Biotechnology
Certificados
Acerca de este artículo
Tumor growth factor alpha (TGF-α) is a member of the epidermal growth factor (EGF) family. TGF-α function is mediated through binding the EGF receptor (EGFR) to activate receptor tyrosine kinase signaling. TGF-α functions as a mitogen to activate epithelial cell proliferation, growth, and differentiation.
- High quality
- Low endotoxin
- Affordable
In the gastric mucosa, TGF-α production inhibits gastric acid secretion and therefore plays a central role in the pathogenesis of the stomach syndrome Ménétrier's disease. TGF-α is also produced in adult macrophages, brain cells, keratinocytes, and is widely expressed in cancer cells.
It is also produced in adult macrophages, brain cells, keratinocytes, and is widely expressed in cancer cells.
Entrega: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Precaución: For research use only.
Especificaciones
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: BALB/c 3T3 proliferation, ED50 ≤2 ng/ml, bioactivity ≥5×10⁵ units/mg
- Gene ID:P01135
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:TGF-alpha|TGFA|EGF-like TGF, ETGF|TGF type 1
- UniProtKB:P01135
- Protein/peptide name:TGF-alpha
- Purity:≥95%
- Molecular weight:5,6 kDa
- Sequence:VVSHFNDCPD SHTQFCFHGT CRFLVQEDKP ACVCHSGYVG ARCEHADLLA
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Especificaciones
Frequently Bought Together