Human Recombinant GRO-A (from E. coli)
Growth regulated protein alpha (GRO-α), also known as CXCL1, is a chemokine that has mitogenic properties and is a neutrophil chemoattractant. GRO-α is secreted by macrophages, epithelial cells, neutrophils, and melanomas. GRO-α signals through the CXCR2 chemokine receptor and is important during spinal cord formation, inflammation, angiogenesis, tumorigenesis, and wound healing.
- High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P09341
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CXCL1|mKC|mgSAα|GRO1|NAP-3|rCINC
- Protein/peptide name:GRO-A
- Purity:≥95%
- Molecular weight:7,9 kDa
- Sequence:ASVATELRCQ CLQTLQGIHP KNIQSVNVKS PGPHCAQTEV IATLKNGRKA CLNPASPIVK KIIEKMLNSD KSN
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Frequently Bought Together