Cobalt(II) fluoride, anhydrous 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10026-17-2
:
Formula:CoF₂
MDL Number:MFCD00010941
:96.93 g/mol
Merck Index:14,2440
Boiling Pt:1400 °C
Melting Pt:1200 °C
Density:4.46
EINECS:233-061-9
UN:2923
ADR:8,III
:GHS06
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together