Cobalt(II) fluoride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10026-17-2
:
Formula:CoF₂
MDL Number:MFCD00010941
:96.93 g/mol
Merck Index:13,02465
Boiling Pt:1400 °C (760 mmHg)
Melting Pt:1200 °C
Density:4,46 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-061-9
UN:2923
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Pink Powder
- Assay:≥98.5 % (ex F)
Specification Test Results
Frequently Bought Together