Order Entry
Spain
ContactUsLinkComponent
Nickel(II) chloride, anhydrous 99.99% (trace metals basis), powder
Toxic
Health
Environment
Nickel(II) chloride, anhydrous 99.99% (trace metals basis), powder
Certificates
undefined
Toxic
Health
Environment
Nickel(II) chloride, anhydrous 99.99% (trace metals basis), powder

Specification Test Results

  • Appearance:
    Yellow to orange Powder
  • Titration Complexometric:
    44.6 to 46.0 % (Ni)
  • Total trace metal impurities:
    ≤100 ppm

Specification Test Results

Related Information

Frequently Bought Together