Human Recombinant WISP-2 (from E. coli)
About this item
WNT1-inducible-signaling pathway protein 2 (WISP-2) is a member of the CYR61/CTGF/NOV (CCN) family of regulatory factors. WISP-2 is expressed in ectodermal, mesodermal, and endodermal lineages, including primary osteoblasts, fibroblasts, mesenchymal stem cells, and adipogenic precursor cells. WISP-2 is a canonical WNT ligand that regulates cell proliferation, adhesion, and metastasis. Secreted WISP-2 promotes mesenchymal precursor cell proliferation and maintains them in an undifferentiated state. In bone-forming osteoblasts, WISP-2 promotes osteoblast adhesion and inhibits osteocalcin production.
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.
5 Options Available Below
Product Details & Documents
WNT1-inducible-signaling pathway protein 2 (WISP-2) is a member of the CYR61/CTGF/NOV (CCN) family of regulatory factors. WISP-2 is expressed in ectodermal, mesodermal, and endodermal lineages, including primary osteoblasts, fibroblasts, mesenchymal stem cells, and adipogenic precursor cells. WISP-2 is a canonical WNT ligand that regulates cell proliferation, adhesion, and metastasis. Secreted WISP-2 promotes mesenchymal precursor cell proliferation and maintains them in an undifferentiated state. In bone-forming osteoblasts, WISP-2 promotes osteoblast adhesion and inhibits osteocalcin production.
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SHBT100-197-5UG
- SHBT100-197-20UG
- SHBT100-197-100UG
- SHBT100-197-500UG
- SHBT100-197-1MG
- Protein/peptide name:
- WISP-2
- WISP-2
- WISP-2
- WISP-2
- WISP-2
- Protein synonyms:
- Connective Tissue Growth Factor-like protein|CT58|CTGF-L|CCN5
- Connective Tissue Growth Factor-like protein|CT58|CTGF-L|CCN5
- Connective Tissue Growth Factor-like protein|CT58|CTGF-L|CCN5
- Connective Tissue Growth Factor-like protein|CT58|CTGF-L|CCN5
- Connective Tissue Growth Factor-like protein|CT58|CTGF-L|CCN5
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- O76076
- O76076
- O76076
- O76076
- O76076
- Sequence:
- MQLCPTPCTC PWPPPRCPLG VPLVLDGCGC CRVCARRLGE PCDQLHVCDA SQGLVCQPGA GPGGRGALCL LAEDDSSCEV NGRLYREGET FQPHCSIRCR CEDGGFTCVP LCSEDVRLPS WDCPHPRRVE VLGKCCPEWV CGQGGGLGTQ PLPAQGPQFS GLVSSLPPGV PCPEWSTAWG PCSTTCGLGM ATRVSNQNRF CRLETQRRLC LSRPCPPSRG RSPQNSAF
- MQLCPTPCTC PWPPPRCPLG VPLVLDGCGC CRVCARRLGE PCDQLHVCDA SQGLVCQPGA GPGGRGALCL LAEDDSSCEV NGRLYREGET FQPHCSIRCR CEDGGFTCVP LCSEDVRLPS WDCPHPRRVE VLGKCCPEWV CGQGGGLGTQ PLPAQGPQFS GLVSSLPPGV PCPEWSTAWG PCSTTCGLGM ATRVSNQNRF CRLETQRRLC LSRPCPPSRG RSPQNSAF
- MQLCPTPCTC PWPPPRCPLG VPLVLDGCGC CRVCARRLGE PCDQLHVCDA SQGLVCQPGA GPGGRGALCL LAEDDSSCEV NGRLYREGET FQPHCSIRCR CEDGGFTCVP LCSEDVRLPS WDCPHPRRVE VLGKCCPEWV CGQGGGLGTQ PLPAQGPQFS GLVSSLPPGV PCPEWSTAWG PCSTTCGLGM ATRVSNQNRF CRLETQRRLC LSRPCPPSRG RSPQNSAF
- MQLCPTPCTC PWPPPRCPLG VPLVLDGCGC CRVCARRLGE PCDQLHVCDA SQGLVCQPGA GPGGRGALCL LAEDDSSCEV NGRLYREGET FQPHCSIRCR CEDGGFTCVP LCSEDVRLPS WDCPHPRRVE VLGKCCPEWV CGQGGGLGTQ PLPAQGPQFS GLVSSLPPGV PCPEWSTAWG PCSTTCGLGM ATRVSNQNRF CRLETQRRLC LSRPCPPSRG RSPQNSAF
- MQLCPTPCTC PWPPPRCPLG VPLVLDGCGC CRVCARRLGE PCDQLHVCDA SQGLVCQPGA GPGGRGALCL LAEDDSSCEV NGRLYREGET FQPHCSIRCR CEDGGFTCVP LCSEDVRLPS WDCPHPRRVE VLGKCCPEWV CGQGGGLGTQ PLPAQGPQFS GLVSSLPPGV PCPEWSTAWG PCSTTCGLGM ATRVSNQNRF CRLETQRRLC LSRPCPPSRG RSPQNSAF
- Protease-free:
- N
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- Molecular weight:
- 24,4 kDa
- 24,4 kDa
- 24,4 kDa
- 24,4 kDa
- 24,4 kDa
- Reconstitution instructions:
- Sterile 10 mM acetic acid at 0.1 mg/ml
- Sterile 10 mM acetic acid at 0.1 mg/ml
- Sterile 10 mM acetic acid at 0.1 mg/ml
- Sterile 10 mM acetic acid at 0.1 mg/ml
- Sterile 10 mM acetic acid at 0.1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SHBT100-197-5UGProtein/peptide nameWISP-2Protein synonymsConnective Tissue Growth Factor-like protein|CT58|CTGF-L|CCN5Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDO76076SequenceMQLCPTPCTC PWPPPRCPLG VPLVLDGCGC CRVCARRLGE PCDQLHVCDA SQGLVCQPGA GPGGRGALCL LAEDDSSCEV NGRLYREGET FQPHCSIRCR CEDGGFTCVP LCSEDVRLPS WDCPHPRRVE VLGKCCPEWV CGQGGGLGTQ PLPAQGPQFS GLVSSLPPGV PCPEWSTAWG PCSTTCGLGM ATRVSNQNRF CRLETQRRLC LSRPCPPSRG RSPQNSAFProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight24,4 kDaReconstitution instructionsSterile 10 mM acetic acid at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT100-197-20UGProtein/peptide nameWISP-2Protein synonymsConnective Tissue Growth Factor-like protein|CT58|CTGF-L|CCN5Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDO76076SequenceMQLCPTPCTC PWPPPRCPLG VPLVLDGCGC CRVCARRLGE PCDQLHVCDA SQGLVCQPGA GPGGRGALCL LAEDDSSCEV NGRLYREGET FQPHCSIRCR CEDGGFTCVP LCSEDVRLPS WDCPHPRRVE VLGKCCPEWV CGQGGGLGTQ PLPAQGPQFS GLVSSLPPGV PCPEWSTAWG PCSTTCGLGM ATRVSNQNRF CRLETQRRLC LSRPCPPSRG RSPQNSAFProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight24,4 kDaReconstitution instructionsSterile 10 mM acetic acid at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT100-197-100UGProtein/peptide nameWISP-2Protein synonymsConnective Tissue Growth Factor-like protein|CT58|CTGF-L|CCN5Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDO76076SequenceMQLCPTPCTC PWPPPRCPLG VPLVLDGCGC CRVCARRLGE PCDQLHVCDA SQGLVCQPGA GPGGRGALCL LAEDDSSCEV NGRLYREGET FQPHCSIRCR CEDGGFTCVP LCSEDVRLPS WDCPHPRRVE VLGKCCPEWV CGQGGGLGTQ PLPAQGPQFS GLVSSLPPGV PCPEWSTAWG PCSTTCGLGM ATRVSNQNRF CRLETQRRLC LSRPCPPSRG RSPQNSAFProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight24,4 kDaReconstitution instructionsSterile 10 mM acetic acid at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT100-197-500UGProtein/peptide nameWISP-2Protein synonymsConnective Tissue Growth Factor-like protein|CT58|CTGF-L|CCN5Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDO76076SequenceMQLCPTPCTC PWPPPRCPLG VPLVLDGCGC CRVCARRLGE PCDQLHVCDA SQGLVCQPGA GPGGRGALCL LAEDDSSCEV NGRLYREGET FQPHCSIRCR CEDGGFTCVP LCSEDVRLPS WDCPHPRRVE VLGKCCPEWV CGQGGGLGTQ PLPAQGPQFS GLVSSLPPGV PCPEWSTAWG PCSTTCGLGM ATRVSNQNRF CRLETQRRLC LSRPCPPSRG RSPQNSAFProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight24,4 kDaReconstitution instructionsSterile 10 mM acetic acid at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRT - Specifications:
More
Specifications
Cat. No.SHBT100-197-1MGProtein/peptide nameWISP-2Protein synonymsConnective Tissue Growth Factor-like protein|CT58|CTGF-L|CCN5Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDO76076SequenceMQLCPTPCTC PWPPPRCPLG VPLVLDGCGC CRVCARRLGE PCDQLHVCDA SQGLVCQPGA GPGGRGALCL LAEDDSSCEV NGRLYREGET FQPHCSIRCR CEDGGFTCVP LCSEDVRLPS WDCPHPRRVE VLGKCCPEWV CGQGGGLGTQ PLPAQGPQFS GLVSSLPPGV PCPEWSTAWG PCSTTCGLGM ATRVSNQNRF CRLETQRRLC LSRPCPPSRG RSPQNSAFProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight24,4 kDaReconstitution instructionsSterile 10 mM acetic acid at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRT



