Dog Recombinant RC GM-CSF (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Granulocyte-macrophage colony-stimulating factor (GM-CSF) is a hematopoietic growth factor produced by endothelial cells, monocytes, fibroblasts, and T cells. GM-CSF stimulates the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells. GM-CSF promotes immune system development and regulates neutrophil function during infection.
- High quality
- Low endotoxin
- Affordable
GM-CSF stimulates the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells.
Delivery: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Dog
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: TF-1 cell proliferation, ED50 ≤15 ng/ml, bioactivity ≥6,7×10⁴ units/mg
- Gene ID:P48749
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:MGI-1GM|CSF-2|Pluripoietin-α
- UniProtKB:P48749
- Protein/peptide name:GM-CSF
- Purity:≥95%
- Molecular weight:14,4 kDa
- Sequence:MAPTRSPTLV TRPSQHVDAI QEALSLLNNS NDVTAVMNKA VKVVSEVFDP EGPTCLETRL QLYKEGLQGS LTSLKNPLTM MANHYKQHCP PTPESPCATQ NINFKSFKEN LKDFLFNIPF DCWKPVKK
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together