Order Entry
Spain
ContactUsLinkComponent
 
Oxidizing
Irritant
Silver iodate 98%
Catalog # 40109.14
CAS Number:  
Oxidizing
Irritant
Silver iodate 98%
Catalog # 40109.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

About this item

 :
 :282.77 g/mol
Merck Index:14,8515
Boiling Pt:1150 °C (1013 hPa)
Melting Pt:>200 °C
Density:5.53
Storage Temperature:Ambient
EINECS:232-039-6
UN:1479
ADR:5.1,II
 :GHS03
 :GHS07
CAS Number:7783-97-3
Formula:AgIO₃
MDL Number:MFCD00014150