Copper(I) cyanide for synthesis, Sigma-Aldrich®
: Merck
:544-92-3
:
:CCuN
:MFCD00010975
:89.56 g/mol
:13,02691
:474 °C
:2.92 g/cm³ (20 °C)
:Ambient
:208-883-6
:6.1,II
:M-Clarity™ quality level = MQ200
:1587
:GHS06
:GHS09
- Solubility:(20 °C) practically insoluble
- Melting Point:473 °C
- Molar Mass:89.56 g/mol
- Bulk Density:750 kg/m3
- Density:2.92 g/cm3
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Frequently Bought Together