
1-Naphthoic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:86-55-5
:
Formula:C₁₁H₈O₂
MDL Number:MFCD00004007
:172.18 g/mol
Merck Index:12,06468
Boiling Pt:300 °C (1013 hPa)
Melting Pt:158…162 °C
Density:1 g/cm³ (20 °C)
EINECS:201-681-9
:GHS07
Specification Test Results
- Appearance:Light yellow to brown Powder
- Infrared spectrum:Conforms
- Melting point:160 to 164 °C
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together