Order Entry
Spain
ContactUsLinkComponent
2-Nitroimidazole 98%
Toxic
2-Nitroimidazole 98%
Catalog # ACRO187570010
CAS Number:  
undefined
Toxic
2-Nitroimidazole 98%
Catalog # ACRO187570010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to khaki Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %
  • Residue after ignition:
    ≤0.1 %

Specification Test Results

About this item

 :
 :113.08 g/mol
Merck Index:13,00923
Melting Pt:287 °C (dec.)
Storage Temperature:Ambient
EINECS:208-425-5
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:527-73-1
Formula:C₃H₃N₃O₂
MDL Number:MFCD00005185