Order Entry
Spain
ContactUsLinkComponent
Dimethyl sulphoxide, dehydrated (max. 0.01% H₂O) ≥99.5%, AnalaR NORMAPUR® analytical reagent
Dimethyl sulphoxide, dehydrated (max. 0.01% H₂O) ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Dimethyl sulphoxide, dehydrated (max. 0.01% H₂O) ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 99.5 %
  • Fe (Iron):
    Max. 0.0001 %
  • Heavy metals (as Pb):
    Max. 0.00010 %
  • Residue on evaporation:
    Max. 0.0010 %
  • Water:
    Max. 0.0100 %

Specification Test Results

Related Information

Frequently Bought Together