
Ammonium iodide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12027-06-4
:
Formula:NH₄I
MDL Number:MFCD00011424
Merck Index:14,529
Melting Pt:551 °C (subl.)
Density:2.514
:144.94 g/mol
EINECS:234-717-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together